2-(4-Aminophenyl)-6-methylbenzothiazole-7-sulphonic acid ≥95% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

CDX-M0118-G010 CDX-M0118-G100
102990-506EA 189.69 USD
102990-506 102990-508
2-(4-Aminophenyl)-6-methylbenzothiazole-7-sulphonic acid ≥95% (by HPLC)
2-(4-Aminophenyl)-6-methylbenzothiazole-7-sulfonic acid

Fluorescent reagent.

Formula: C₁₄H₁₂N₂O₃S₂
Storage Temperature: Refrigerator
CAS Number: 130-17-6

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (HPLC)
Appearance/Color Solid.
Solubility Soluble in acetonitrile or DMSO.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR