H-Met-AMC ≥99% (by HPLC, sum of enantiomers)
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
L-Methionine 7-amido-4-methylcoumarin trifluoroacetate salt
CDX-M0113-M100
CDX-M0113-G002
102990-498EA
202.15
USD
102990-498
102990-500
H-Met-AMC ≥99% (by HPLC, sum of enantiomers)
H-Met-AMC
Substrate for calpain.
Formula:
C₁₅H₁₈N₂O₃S·C₂HF₃O₂ MW: 420.40 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00210065 CAS Number: 94367-35-8 |
Specification Test Results
Source/Host | Synthetic. |
Purity | >99% (HPLC, sum of enantiomers) |
Appearance/Color | Solid. |
Solubility | Soluble in DMSO. |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...