2′-(4-Methylumbelliferyl)-alpha-D-N-acetylneuraminic acid sodium salt hydrate ≥95% (by NMR)

Supplier: ADIPOGEN CORP MS

Synonyms: 4-MUNANA

CDX-M0096-M001 CDX-M0096-M005 CDX-M0096-M025
102990-486EA 70.62 USD
102990-486 102990-488 102990-490
2′-(4-Methylumbelliferyl)-alpha-D-N-acetylneuraminic acid sodium salt hydrate ≥95% (by NMR)
2′-(4-Methylumbelliferyl)-alpha-D-N-acetylneuraminic acid sodium salt hydrate

Fluorogenic substrate used to measure sensitively enzymatic neuraminidase activity in fluorometric assays in mammalian and viral sources. Neuraminidase is catalyzed by the hydrolysation to form N-acetylneuraminic acid and the blue fluorescent soluble compound 4-Methylumbelliferone, which can be detected fluorometrically. Spectral data: lambdaex 315 nm| lambdaem 374 nm (pH.7.0) or lambdaex 365 nm| lambdaem 445 nm (after cleavage by neuraminidase). Used for fluorescent staining of sialidases in PAGE.

Formula: C₂₁H₂₄NNaO₁₁·xH₂O
MW: 489.41 g/mol (anhy)
Storage Temperature: Freezer
MDL Number: MFCD00057309
CAS Number: 76204-02-9

Order Now

Specification Test Results

Source/Host Synthetic
Purity 95% (NMR)
Appearance/Color Light yellow powder.
Solubility Soluble in water or methanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR