4-Methylumbelliferyl oleate ≥95%

Supplier: ADIPOGEN CORP MS

Synonyms: 4-Methyl-2-oxo-2H-chromen-7-yl 9-octadecenoate

CDX-M0086-M025 CDX-M0086-M250 CDX-M0086-GG25
102990-470EA 84.46 USD
102990-470 102990-472 102990-474
4-Methylumbelliferyl oleate ≥95%
4-Methylumbelliferyl oleate

Fluorogenic substrate for lipases, which can be used in aqueous solutions as well as several organic solvents.

Formula: C₂₈H₄₀O₄
MW: 440.61 g/mol
Storage Temperature: Refrigerator
CAS Number: 18323-58-5

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (HPCE)
Appearance/Color White to off-white solidified mass.
Solubility Soluble in 2-Methoxyethanol, DMSO or chloroform.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR