7-Methylguanine ≥98%

Supplier: ADIPOGEN CORP MS

Synonyms: 2-Amino-7-methyl-1H-purin-6(7H)-one, N7-Methylguanine

CDX-M0082-M250 CDX-M0082-M500 CDX-M0082-G001
102990-464EA 139.85 USD
102990-464 102990-466 102990-468
7-Methylguanine ≥98%
7-Methylguanine

Integral part of the cap structure of mRNA necessary for the attachment of the nucleic acids to the ribosome. Has binding affinity to guanine deaminase, an important metalloenzyme in purine catabolism. Found in urine of patients with malignant cancer, suggesting an important role.

Formula: C₆H₇N₅O
MW: 165.15 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00038003
CAS Number: 578-76-7

Order Now

Specification Test Results

Source/Host Synthetic
Purity 98% (HPLC)
Appearance/Color White powder.
Solubility Soluble in DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR