N-(N'-Maleinimidyl-2-ethyl)-4-(2-(6-(3,4-dihydro-2H-1-benzopyranyl))-5-oxyzolyl) pyridinium triflate ≥90% (by HPCE)

Supplier: ADIPOGEN CORP MS
Warning

CDX-M0081-M005 CDX-M0081-M050 CDX-M0081-M250
102990-450EA 58.15 USD
102990-450 102990-452 102990-462
N-(N'-Maleinimidyl-2-ethyl)-4-(2-(6-(3,4-dihydro-2H-1-benzopyranyl))-5-oxyzolyl) pyridinium triflate ≥90% (by HPCE)
N-(N'-Maleinimidyl-2-ethyl)-4-(2-(6-(3,4-dihydro-2H-1-benzopyranyl))-5-oxyzolyl)pyridiniumtriflate

Highly photostable, water soluble fluorescent label with large Stoke′s shift, high QY in aqueous media and pH tolerance.

Formula: C₂₄H₂₀F₃N₃O₇S
MW: 551.49 g/mol
Storage Temperature: Freezer
CAS Number: 155863-05-1

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 90% (HPCE)
Appearance/Color Solid.
Solubility Soluble in DMSO.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR