1-[2-(Maleimido)ethyl]-4-[5-(4-methoxyphenyl)-2-oxazolyl]pyridinium triflate ≥90%

Supplier: ADIPOGEN CORP MS
Warning

CDX-M0080-M005 CDX-M0080-M050 CDX-M0080-M250
102990-444EA 58.15 USD
102990-444 102990-446 102990-448
1-[2-(Maleimido)ethyl]-4-[5-(4-methoxyphenyl)-2-oxazolyl]pyridinium triflate ≥90%
1-[2-(Maleimido)ethyl]-4-[5-(4-methoxyphenyl)-2-oxazolyl]pyridinium triflate

Highly photostable, water soluble fluorescent label with large Stoke′s shift, high QY in aqueous media and pH tolerance.

Formula: C₂₂H₁₈F₃N₃O₇S
Storage Temperature: Freezer
CAS Number: 155862-98-9

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 90% (HPCE)
Appearance/Color Solid.
Solubility Soluble in DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR