2-Maleinimidoethyl mesylate ≥90% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: Linker 8

CDX-M0078-G001 CDX-M0078-GG25 CDX-M0078-G010
102990-438EA 325.38 USD
102990-438 102990-440 102990-442
2-Maleinimidoethyl mesylate ≥90% (by HPLC)
2-Maleinimidoethyl mesylate

Building block for synthesis.

Formula: C₇H₉NO₅S
MW: 219.22 g/mol
Storage Temperature: Freezer
CAS Number: 155863-36-8

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 90% (HPLC)
Appearance/Color Off-white powder.
Solubility Soluble in chloroform.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR