MPTS ≥98% (by TLC)

Supplier: ADIPOGEN CORP MS

CDX-M0055-M100 CDX-M0055-M250 CDX-M0055-G001
102990-420EA 163.38 USD
102990-420 102990-422 102990-424
MPTS ≥98% (by TLC)
MPTS

Highly water-soluble superpolar fluorescent probe for assaying cationic surfactants. Can be used as a polar tracer. It has also been used in combination with a boronic acid-modified viologen quencher to sense glucose at pH 7.4 in buffered aqueous solution.

Formula: C₁₇H₉Na₃O₁₀S₃
MW: 538.41 g/mol
Storage Temperature: Freezer
CAS Number: 82962-86-5

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (TLC)
Appearance/Color Light brown to beige crystalline powder.
Solubility Soluble in water, methanol, DMSO or DMF.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR