5-Methoxy-2,3,3-trimethyl-1-(4-sulfobutyl)indolium inner salt ≥95% (by HPCE)

Supplier: ADIPOGEN CORP MS
Warning

CDX-M0049-M500 CDX-M0049-GG25
102990-416EA 181.38 USD
102990-416 102990-418
5-Methoxy-2,3,3-trimethyl-1-(4-sulfobutyl)indolium inner salt ≥95% (by HPCE)
5-Methoxy-2,3,3-trimethyl-1-(4-sulfobutyl)indolium inner salt

Fluorescent reagent.

Formula: C₁₆H₂₃NO₄S
MW: 325.42 g/mol
Storage Temperature: Freezer
CAS Number: 54136-27-5

Order Now

Specification Test Results

Source/Host Synthetic.
Purity >95% (HPCE)
Appearance/Color Solid.
Solubility Soluble in DMSO.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR