Merocyanin 540 ≥90%
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
Sodium 3-(2-(4-(1,3-dibutyl-4,6-dioxo-2-thioxotetrahydropyrimidin-5(2H)-ylidene)but-2-en-1-ylidene)benzo[d]oxazol-3(2H)-yl)propane-1-sulphonate
CDX-M0033-M100
CDX-M0033-M500
CDX-M0033-G005
102990-406EA
123.23
USD
102990-406
102990-408
102990-410
Merocyanin 540 ≥90%
Merocyanin 540
Sensitive probe for membrane potential (Ex/Em: 555/578nm in methanol). Selective staining of immature hemopoietic cells in flow cytometry.
Formula:
C₂₆H₃₂N₃NaO₆S₂ MW: 569.67 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00013438 CAS Number: 62796-23-0 |
Specification Test Results
Source/Host | Synthetic. |
Purity | 90% (Dye Content) |
Appearance/Color | Blue solid. |
Solubility | Soluble in water, DMSO, ethanol or methanol. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...