7-Methoxy-4-coumarinylacetic acid N-succinimidyl ester ≥90% (by NMR)

Supplier: ADIPOGEN CORP MS

CDX-M0019-M050 CDX-M0019-M250
102990-386EA 180 USD
102990-386 102990-388
7-Methoxy-4-coumarinylacetic acid N-succinimidyl ester ≥90% (by NMR)
7-Methoxy-4-coumarinylacetic acid N-succinimidyl ester

Fluorescent marker for peptides.

Formula: C₁₆H₁₃NO₇
Storage Temperature: Freezer
CAS Number: 359436-89-8

Order Now

Specification Test Results

Source/Host Synthetic.
Purity >90% (NMR)
Appearance/Color Solid.
Solubility Soluble in acetonitrile.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR