7-Methoxycoumarin-3-carbonyl azide ≥97% (by HPCE)

Supplier: ADIPOGEN CORP MS
Warning

CDX-M0018-M100 CDX-M0018-M500 CDX-M0018-G001
102990-380EA 130.15 USD
102990-380 102990-382 102990-384
7-Methoxycoumarin-3-carbonyl azide ≥97% (by HPCE)
7-Methoxycoumarin-3-carbonyl azide

Fluorescent reagent.

Formula: C₁₁H₇N₃O₄
Storage Temperature: Freezer
CAS Number: 97632-67-2

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 97% (HPCE)
Appearance/Color Yellow powder.
Solubility Soluble in DMSO, DMF or acetonitrile.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR