7-Methoxy-4-(trifluoromethyl)coumarin ≥99% (by TLC)

Supplier: ADIPOGEN CORP MS
Warning

CDX-M0015-M050 CDX-M0015-M250
102990-368EA 202.15 USD
102990-368 102990-370
7-Methoxy-4-(trifluoromethyl)coumarin ≥99% (by TLC)
7-Methoxy-4-(trifluoromethyl)coumarin

Fluorescent substrate for oxidoreductases, especially for Cytochrome P450.

Formula: C₁₁H₇F₃O₃
Storage Temperature: Freezer
MDL Number: MFCD00132980
CAS Number: 575-04-2

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 99% (TLC)
Appearance/Color White powder.
Solubility Soluble in ethanol, methanol, DMSO or DMF.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR