5-Maleimidoeosin ≥93%

Supplier: ADIPOGEN CORP MS

Synonyms: Eosin-5-maleimide

CDX-M0013-M010 CDX-M0013-M100
102990-364EA 271.38 USD
102990-364 102990-366
5-Maleimidoeosin ≥93%
5-Maleimidoeosin

Triplet probe used for the measurment of rotational diffusion of protein in solution and in membranes, includin myosin and pyruvate dehydrogenase.

Formula: C₂₄H₉Br₄NO₇
Storage Temperature: Freezer
CAS Number: 150322-02-4

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 93% (HPLC)
Appearance/Color Solid.
Solubility Soluble in water.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR