NBD-methylhydrazine ≥97% (by HPLC)
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
4-(1-Methylhydrazino)-7-nitrobenzofurazan
N-methyl-4-hydrazino-7-nitrobenzofurezan (MNBDH) is a new reagent for the determination of aldehydes and ketones. MNBDH reacts with carbonyl copmounds in acidic media to the corresponding MNBD-hydrazones. For example formaldehyde hydrazone formed is determined by high-performance liquid chromatography with UV/visible detection at 474 nm. MNBDH has been studied recently as a reagent with reduced interferences by ozone and nitrogen dioxide. It reacts with these oxidants to give only one product N-methyl-4-amino-7-nitrobenzofurezan (MNBDA).
Formula:
C₇H₇N₅O₃ MW: 209.16 g/mol Storage Temperature: Freezer |
CAS Number:
214147-22-5 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in chloroform. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...