Lithium tetrakis(pentafluorophenyl)borate ethyl etherate ≥97% (by F-NMR)

Supplier: ADIPOGEN CORP MS
Warning

CDX-L0011-M100 CDX-L0011-G001 CDX-L0011-G010
102990-324EA 91.38 USD
102990-324 102990-326 102990-328
Lithium tetrakis(pentafluorophenyl)borate ethyl etherate ≥97% (by F-NMR)
Lithium tetrakis(pentafluorophenyl)borate ethyl etherate

Salt for the generation of cationic transition metal complexes.

Formula: C₂₄BF₂₀Li·2.5C₄H₁₀O
MW: 871.28 g/mol
Storage Temperature: Freezer
CAS Number: 371162-53-7

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 97% (F-NMR)
Appearance/Color Beige powder.
Solubility Soluble in dichloromethane, toluene or acetonitrile.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR