Bis(4-iodophenyl)-iodonium tetrafluoroborate ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS

CDX-I0053-G001 CDX-I0053-G005
102990-304EA 175.85 USD
102990-304 102990-306
Bis(4-iodophenyl)-iodonium tetrafluoroborate ≥98% (by HPLC)
Bis(4-iodophenyl)iodonium tetrafluoroborate

Diaryliodonium salts. Use for the synthesis of tri-, tetra-, and penta-substituted pyrrole compounds.

Formula: C₁₂H₈I₃·BF₄
MW: 619.7 g/mol
Storage Temperature: Freezer
CAS Number: 1260147-20-3

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (HPLC)
Appearance/Color White powder.
Solubility Soluble in chloroform.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR