Isoprothiolane ≥98% (by HPLC)
Supplier: ADIPOGEN CORP MS
|
Warning
|
Isoprothiolane is a dithiolane pesticide. Isoprothiolane is commonly used in agriculture as a fungicide and insecticide to control planthoppers and blast disease in rice plants. Shown to regulate mammary epithelial cell growth in vitro by modulation production of cytokines and improving hepatic lipid metabolism in cows. Compound can be used as analytical reference material.
Formula:
C₁₂H₁₈O₄S₂ Storage Temperature: Freezer |
MDL Number:
MFCD00210314 CAS Number: 50512-35-1 UN: 3077 ADR: 9,III |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | White to light yellow/beige powder. |
Solubility | Soluble in chloroform. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...