1(E)-Iodo-hex-1-ene ≥97% (by GC)

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: (E)-1-Iodohex-1-ene

CDX-I0047-G001 CDX-I0047-G005
102990-292EA 105.23 USD
102990-292 102990-294
1(E)-Iodo-hex-1-ene ≥97% (by GC)
1(E)-Iodohex-1-ene

Building block for synthesis.

Formula: C₆H₁₁I
MW: 210.1 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00798209
CAS Number: 16644-98-7
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 97% (GC)
Appearance/Color Brown liquid.
Solubility Soluble in chloroform.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR