1-[2-(4-Isothiocyanatophenoxy)ethyl]-4-[5-(4-methoxyphenyl)-2-oxazolyl] pyridinium tosylate ≥95% (by HPCE)

Supplier: ADIPOGEN CORP MS
Danger

CDX-I0033-M025 CDX-I0033-M050 CDX-I0033-M250
102990-186EA 141.23 USD
102990-186 102990-188 102990-190
1-[2-(4-Isothiocyanatophenoxy)ethyl]-4-[5-(4-methoxyphenyl)-2-oxazolyl] pyridinium tosylate ≥95% (by HPCE)
1-[2-(4-Isothiocyanatophenoxy)ethyl]-4-[5-(4-methoxyphenyl)-2-oxazolyl]pyridinium tosylate

Class of highly photostable, water soluble fluorescent labels with large Stoke′s shift, high QY in aqueous media and pH tolerance. Relatively low extinction coefficient (Epsilon∼ 20,000 in EtOH).

Formula: C₃₁H₂₇N₃O₆S₂
Storage Temperature: Freezer
CAS Number: 155862-93-4
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (HPCE)
Appearance/Color Solid.
Solubility Soluble in chloroform.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR