2-Iodoxy-3,4,5,6-tetrafluorobenzoic acid ≥97% (by NMR)
Supplier: ADIPOGEN CORP MS
Synonyms:
FIBX
CDX-I0028-M500
CDX-I0028-GG25
102990-176EA
210.46
USD
102990-176
102990-178
2-Iodoxy-3,4,5,6-tetrafluorobenzoic acid ≥97% (by NMR)
2-Iodoxy-3,4,5,6-tetrafluorobenzoic acid
Convenient substitute of o-iodoxybenzoic acid (IBX) as an oxidant. FIBX is safer, has neutral properties behaves as an equivalent to IBX in the oxidation of alcohols to ketones or aldehydes in polar solvents and shows better solubility.
Formula:
C₇HF₄IO₄ MW: 352 g/mol Storage Temperature: Freezer |
CAS Number:
954373-95-6 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | White to off-white solid. |
Solubility | Soluble in toluene. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...