Indo-1 pentapotassium salt ≥90%

Supplier: ADIPOGEN CORP MS

Synonyms: Indo-1 5K⁺

CDX-I0018-M002 CDX-I0018-M025
102990-160EA 263.08 USD
102990-160 102990-162
Indo-1 pentapotassium salt ≥90%
Indo-1 pentapotassium salt

Fluorescent calcium chelator and indicator with an unusually large shift in fluorescence emission from 480 nm to 400 nm on binding of calcium. Useful for intracellular application.

Formula: C₃₂H₂₆K₅N₃O₁₂
Storage Temperature: Freezer
CAS Number: 132319-56-3

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 90% (HPLC)
Appearance/Color Off-white solid.
Solubility Soluble in water.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR