Resorufinisobutyrate ≥97%
Supplier: ADIPOGEN CORP MS
Synonyms:
7-(Isobutyloxycarbonyloxy)-3H-phenoxazin-3-one
CDX-I0005-M001
CDX-I0005-M005
CDX-I0005-M025
102990-148EA
137.08
USD
102990-148
102990-150
102990-152
Resorufinisobutyrate ≥97%
Resorufinisobutyrate
Cell permeable resorufin derivative used as a fluorogenic indicator for cell viability. Resorufin was selected as the fluorophore because it fluoresces at a longer wavelength and is amenable to modification at the hydroxyl group which is essential for its fluorescence. Incubation with esterase at pH 8.0 results in a 80-90 nm shift of emission max. (Ex/Em: 500/~593nm in 0.1 M Tris pH 8.0 (after cleavage by esterase)). This cell viability indicator is 2 times more sensitive than calcein-AM.
Formula:
C₁₇H₁₅NO₅ Storage Temperature: Freezer |
CAS Number:
251292-24-7 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMSO. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...