N-Heptanoyl-DL-homoserine lactone ≥97% (by GC)
Supplier: ADIPOGEN CORP MS
Synonyms:
C7-HSL
CDX-H0201-M010
CDX-H0201-M050
102990-136EA
88.62
USD
102990-136
102990-138
N-Heptanoyl-DL-homoserine lactone ≥97% (by GC)
N-Heptanoyl-DL-homoserine lactone
N-Heptanoyl-L-homoserine lactone (C7-HSL) is a small diffusible signaling molecule involved in quorum sensing, thereby controlling gene expression and affecting cellular metabolism. The diverse applications of this kind of molecule include regulation of virulence, infection prevention and septicemia in fish.
Formula:
C₁₁H₁₉NO₃ MW: 213.3 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD01862914 CAS Number: 106983-26-0 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | White solid. |
Solubility | Soluble in chloroform. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...