(R)-3-Hydroxymyristic acid ≥98% (by NMR)

Supplier: ADIPOGEN CORP MS

CDX-H0114-M250 CDX-H0114-G001
102990-114EA 508.15 USD
102990-114 102990-116
(R)-3-Hydroxymyristic acid ≥98% (by NMR)
(R)-3-Hydroxymyristic acid

Hydroxy fatty acid used to study its role in biological processes such as oxidative stress, inflammation and insulin resistance. Used in endotoxin and lipid A research, since it is one of two major components of bacterial Lipid A. Shows antifungal activity. Compound could be used as analytical reference in assaying food samples.

Formula: C₁₄H₂₈O₃
Storage Temperature: Freezer
MDL Number: MFCD00211333
CAS Number: 28715-21-1

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (NMR)
Appearance/Color White powder.
Solubility Soluble in chloroform, dichloromethane, ether or ethyl acetate.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR