N-Hexadecanoyl-L-homoserine lactone ≥97% (by GC)

Supplier: ADIPOGEN CORP MS

Synonyms: (S)-N-(2-Oxotetrahydrofuran-3-yl)palmitamide, C16-HSL

CDX-H0113-M010 CDX-H0113-M050
102990-110EA 88.62 USD
102990-110 102990-112
N-Hexadecanoyl-L-homoserine lactone ≥97% (by GC)
N-Hexadecanoyl-L-homoserine lactone

N-Hexadecanoyl-L-homoserine lactone is a small diffusible signaling molecule and is a member of N-acyl-homoserine lactone family. N-acylhomoserine lactones (AHL) are involved in quorum sensing, controlling gene expression, and cellular metabolism. C16-HSL is a lipophilic, long acyl side-chain bearing AHL, produced by the LuxI AHL synthase homolog SinI involved in quorum-sensing signaling in S. meliloti, a nitrogen-fixing bacterial symbiont of certain legumes. C16-HSL is the most abundant AHL produced by the proteobacterium R. capsulatus and activates genetic exchange between R. capsulatus cells. It tends to localize in relatively lipophilic cellular environments of bacteria and cannot diffuse freely through the cell membrane. The long-chain N-acylhomoserine lactones may be exported from cells by efflux pumps or may be transported between communicating cells by extracellular outer membrane vesicles.

Formula: C₂₀H₃₇NO₃
MW: 339.5 g/mol
Storage Temperature: Freezer
MDL Number: MFCD18382092
CAS Number: 87206-01-7

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 97% (GC)
Appearance/Color White solid.
Solubility Soluble in chloroform.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR