N-Heptanoyl-L-homoserine lactone ≥98% (by NMR)

Supplier: ADIPOGEN CORP MS

Synonyms: N-[(3S)-Tetrahydro-2-oxo-3-furanyl]-heptanamide

CDX-H0112-M005 CDX-H0112-M050
102990-106EA 88.62 USD
102990-106 102990-108
N-Heptanoyl-L-homoserine lactone ≥98% (by NMR)
N-Heptanoyl-L-homoserine lactone

N-Heptanoyl-L-homoserine lactone (C7-HSL) is a small diffusible signaling molecule involved in quorum sensing, thereby controlling gene expression and affecting cellular metabolism. The diverse applications of this molecule include regulation of virulence, infection prevention and septicemia in fish.

Formula: C₁₁H₁₉NO₃
MW: 213.3 g/mol
Storage Temperature: Refrigerator
CAS Number: 177158-20-2

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (NMR)
Appearance/Color White powder.
Solubility Soluble in DMSO and dimethylformamide (approx. 30mg/ml).

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR