N-Hexanoyl-L-homoserine lactone ≥98% (by NMR)
Supplier: ADIPOGEN CORP MS
Synonyms:
(S)-N-(2-Oxotetrahydrofuran-3-yl)hexanamide, N-[(3S)-Tetrahydro-2-oxo-3-furanyl]-hexanamide
CDX-H0111-M010
CDX-H0111-M050
102990-102EA
88.62
USD
102990-102
102990-104
N-Hexanoyl-L-homoserine lactone ≥98% (by NMR)
N-Hexanoyl-L-homoserine lactone
N-Hexanoyl-L-homoserine lactone is a small diffusible signaling molecule involved in quorum sensing, controlling gene expression and affecting cellular metabolism. The diverse applications of this molecule include regulation of virulence in general and in cystic fibrosis, infection prevention, slime and biofilm reduction in commercial agriculture and aquaculture industries, food spoilage prevention and septicemia in fish.
Formula:
C₁₀H₁₇NO₃ MW: 199.3 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD12912260 CAS Number: 147852-83-3 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | White powder. |
Solubility | Soluble in DMSO and dimethylformamide (approx. 30mg/ml). |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...