Hydramethylnon ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS
Danger

Synonyms: AC 217,300

CDX-H0096-M100 CDX-H0096-G001
102990-070EA 131.54 USD
102990-070 102990-072
Hydramethylnon ≥98% (by HPLC)
Hydramethylnon

Insecticide primarily used against cockroaches and ants. Chemical class called trifluoromethyl aminohydrazone, which is a metabolic inhibitor. It works by inhibiting complex III in the mitochondrial inner membrane and leads to a halting of oxidative phosphorylation. Compound can be used as analytical reference material.

Formula: C₂₅H₂₄F₆N₄
MW: 494.5 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00128045
CAS Number: 67485-29-4
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (HPLC)
Appearance/Color White to dark yellow powder.
Solubility Soluble in chloroform.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR