DL-3-Hydroxyisobutyric acid sodium salt ≥96% (by GC)

Supplier: ADIPOGEN CORP MS

Synonyms: (±)-Sodium beta-hydroxyisobutyrate

CDX-H0085-M250 CDX-H0085-G001
102990-062EA 123.23 USD
102990-062 102990-064
DL-3-Hydroxyisobutyric acid sodium salt ≥96% (by GC)
DL-3-Hydroxyisobutyric acid sodium salt

Intermediate formed in the valine catabolism. It is a reactant of the enzymes involved in branched chain amino acid metabolism 3-hydroxyisobutyrate dehydrogenase and 3-hydroxyisobutyryl-CoA hydrolase and it was found to have increased concentrations in MS patient metabolic profiles. This compound may be used to study the distribution, characterization and kinetics of these enzymes. Shown to inhibit key enzymes of the energy metabolism.

Formula: C₄H₇NaO₃
MW: 126.09 g/mol
Storage Temperature: Freezer
MDL Number: MFCD16875405
CAS Number: 1219589-99-7

Order Now

Specification Test Results

Source/Host Synthetic
Purity 96% (GC)
Appearance/Color White to off-white powder.
Solubility Soluble in DMSO.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR