3-Hydroxy-hexanoyl-DL-homoserine lactone ≥98% (by NMR)

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: 3OH-C6-HSL RS/RS

CDX-H0082-M005 CDX-H0082-M025
102990-274EA 123.23 USD
102990-274 102990-276
3-Hydroxy-hexanoyl-DL-homoserine lactone ≥98% (by NMR)
3-Hydroxyhexanoyl-DL-homoserine lactone

3-Hydroxy-hexanoyl-DL-homoserine lactone is a small diffusible signaling molecule and is a member of N-acyl-homoserine lactone family. N-acylhomoserine lactones (AHL) are involved in quorum sensing, controlling gene expression, and cellular metabolism. The diverse applications of this kind of molecule include regulation of virulence in general, infection prevention, and formation of biofilms.

Formula: C₁₀H₁₇NO₄
MW: 215.25 g/mol
Storage Temperature: Freezer
CAS Number: 161234-45-3

Order Now

Specification Test Results

Source/Host Synthetic
Purity 98% (NMR)
Appearance/Color White to pink powder.
Solubility Soluble in ethanol, methanol, DMF or DMSO. Poorly soluble in water.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR