N-(3-Hydroxydodecanoyl)-DL-homoserine lactone 97

Supplier: ADIPOGEN CORP MS

CDX-H0081-M025 CDX-H0081-M250
102990-270EA 193.85 USD
102990-270 102990-272
N-(3-Hydroxydodecanoyl)-DL-homoserine lactone 97
N-(3-Hydroxydodecanoyl)-DL-homoserine lactone

Quorum sensing agent.

Formula: C₁₆H₂₉NO₄
MW: 299.41 g/mol
Storage Temperature: Freezer
CAS Number: 182359-60-0

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 97% (HPLC)
Appearance/Color Solid.
Solubility Soluble in DMF or acetonitrile.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR