N-(3-Hydroxydecenoyl)-DL-homoserine lactone ≥99%

Supplier: ADIPOGEN CORP MS

Synonyms: 3-Hydroxy-N-(tetrahydro-2-oxo-3-furanyl)-decanamide

CDX-H0079-M025 CDX-H0079-M250
102990-262EA 315.69 USD
102990-262 102990-264
N-(3-Hydroxydecenoyl)-DL-homoserine lactone ≥99%
N-(3-Hydroxydecenoyl)-DL-homoserine lactone

Quorum sensing agent.

Formula: C₁₄H₂₅NO₄
MW: 271.3558 g/mol
Storage Temperature: Freezer
CAS Number: 929222-14-0

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 99% (HPLC)
Appearance/Color Solid.
Solubility Soluble in DMF or acetonitrile.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR