N-(3-Hydroxyoctanoyl)-DL-homoserine lactone ≥99%

Supplier: ADIPOGEN CORP MS

Synonyms: 3-Hydroxy-N-(tetrahydro-2-oxo-3-furanyl)-octanamide

CDX-H0078-M025 CDX-H0078-M250
102990-258EA 307.38 USD
102990-258 102990-260
N-(3-Hydroxyoctanoyl)-DL-homoserine lactone ≥99%
N-(3-Hydroxyoctanoyl)-DL-homoserine lactone

Quorum sensing agent.

Formula: C₁₂H₂₁NO₄
MW: 243.3022 g/mol
Storage Temperature: Freezer
CAS Number: 853799-77-6

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 99% (HPLC)
Appearance/Color Solid.
Solubility Soluble in DMF or acetonitrile.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR