3-Hydroxy-butanoyl-DL-homoserine lactone ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS

Synonyms: 3-Hydroxy-C4-HSL

CDX-H0076-M005 CDX-H0076-M010
102990-250EA 210.46 USD
102990-250 102990-252
3-Hydroxy-butanoyl-DL-homoserine lactone ≥98% (by HPLC)
3-Hydroxybutanoyl-DL-homoserine lactone

3-Hydroxy-butanoyl-DL-homoserine lactone is a small diffusible signaling molecule and is a member of N-acyl-homoserine lactone family. N-acylhomoserine lactones (AHL) are involved in quorum sensing, controlling gene expression, and cellular metabolism. The diverse applications of this kind of molecule include regulation of virulence in general, infection prevention, and formation of biofilms.

Formula: C₈H₁₃NO₄
MW: 187.19 g/mol
Storage Temperature: Freezer
CAS Number: 912545-51-8

Order Now

Specification Test Results

Source/Host Synthetic
Purity 98% (HPLC)
Appearance/Color White powder.
Solubility Soluble in chloroform.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR