N-(3-Hydroxytetradecanoyl)-DL-homoserine lactone ≥96% (by HPLC)
Supplier: ADIPOGEN CORP MS
|
Danger
|
CDX-H0044-M020
CDX-H0044-M250
102990-240EA
210.46
USD
102990-240
102990-242
N-(3-Hydroxytetradecanoyl)-DL-homoserine lactone ≥96% (by HPLC)
N-(3-Hydroxytetradecanoyl)-DL-homoserine lactone
Quorum sensing agent.
Formula:
C₁₈H₃₃NO₄ Storage Temperature: Freezer |
CAS Number:
172670-99-4 UN: 2811 ADR: 6.1,III |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMF or acetonitrile. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...