BHHCT ≥90% (by HPCE)

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: 4,4''-Bis(4,4,5,5,6,6,6-heptafluoro-3-oxohexanoyl)-[1,1':2',1''-terphenyl]-4'-sulphonyl chloride, 4,4′′-Bis(4,4,5,5,6,6,6-heptafluoro-1,3-dioxohexyl)-o-terphenyl-4′-sulphonyl chloride

CDX-H0041-M005 CDX-H0041-M100 CDX-H0041-G001
102990-230EA 91.38 USD
102990-230 102990-232 102990-234
BHHCT ≥90% (by HPCE)
BHHCT

Sensitive label for time-resolved fluoroimmunoassay via the europium chelate. Ligand for TRF-complexes (with Eu3+) used for preparing Silica-Nanoparticles. This label is characterized by very large Stoke's shift, broad exitation and narrow emission bands.

Formula: C₃₀H₁₅ClF₁₄O₆S
MW: 804.93 g/mol
Storage Temperature: Freezer
MDL Number: MFCD21609376
CAS Number: 200862-70-0

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 90% (HPCE)
Appearance/Color Pink powder.
Solubility Soluble in DMSO.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR