Phosphazenium tetrafluoroborate P2-BF4 ≥98% (by titrimetric analysis)
Supplier: ADIPOGEN CORP MS
|
Danger
|
Synonyms:
1,1,1,3,3,3-Hexakis(dimethylamino)diphosphazenium tetrafluoroborate
CDX-H0011-G001
CDX-H0011-G005
CDX-H0011-G025
102990-208EA
88.62
USD
102990-208
102990-210
102990-212
Phosphazenium tetrafluoroborate P2-BF4 ≥98% (by titrimetric analysis)
Phosphazenium tetrafluoroborate P2-BF4
A phase transfer catalyst directing enolate alkylation towards O-substitution. Starting material for preparing the corresponding fluoride, a very reactive naked fluoride, with KF.
Formula:
C₁₂H₃₆BF₄N₇P₂ MW: 427.21 g/mol Storage Temperature: Freezer |
CAS Number:
137334-98-6 |
Specification Test Results
Source/Host | Synthetic |
Purity |
|
Appearance/Color | Colourless crystalline. |
Solubility | Soluble in THF. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...