7-Hydroxy-4-methyl-2(1H)-quinolone ≥97%

Supplier: ADIPOGEN CORP MS
Warning

CDX-H0010-M200 CDX-H0010-G001
102990-060EA 263.08 USD
102990-060 102990-206
7-Hydroxy-4-methyl-2(1H)-quinolone ≥97%
7-Hydroxy-4-methyl-2(1H)-quinolone

7-Hydroxy-4-methyl-2(1H)-quinolone is a starting material for the synthesis of a potential cytotoxic compound class with activity against human tumor cell lines.

Formula: C₁₀H₉NO₂
Storage Temperature: Freezer
CAS Number: 20513-71-7

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 97% (HPLC)
Appearance/Color Solid.
Solubility Soluble in DMSO, DMF or alcohols.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR