HCPI ≥90% (by NMR)

Supplier: ADIPOGEN CORP MS
Danger

Synonyms: 2-[4-(Hydrazinocarbonyl)phenyl]-4,5-diphenylimidazole

CDX-H0003-M050 CDX-H0003-M250
102990-056EA 178.62 USD
102990-056 102990-058
HCPI ≥90% (by NMR)
HCPI

2-(4-Hydrazinocarbonylphenyl)-4,5-diphenylimidazole (HCPI) as a lophine derivative is a versatile fluorescent derivatization reagent for fatty acids. HCPI has a hydrazinocarbonyl group which reacts with fatty acids resulting in intensely fluorescent derivatives.

Formula: C₂₂H₁₈N₄O
MW: 354.4 g/mol
Storage Temperature: Freezer
CAS Number: 151589-38-7
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 90% (NMR)
Appearance/Color Solid.
Solubility Soluble in DMF.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR