D-Fructose-1,6-diphosphate trisodium salt, hydrate ≥98%

Supplier: ADIPOGEN CORP MS

Synonyms: D(+)-Fructofuranose 1,6-diphosphate trisodium salt hydrate, Hexose diphosphate trisodium salt

CDX-F0218-G001 CDX-F0218-G005 CDX-F0218-G050
102990-032EA 78.92 USD
102990-032 102990-034 102990-036
D-Fructose-1,6-diphosphate trisodium salt, hydrate ≥98%
D-Fructose-1,6-diphosphate trisodium salt

D-Fructose-1,6-bisphosphate (FBP), a common metabolic sugar, is the precursor of glyceraldehyde 3-phosphate and dihydroxyacetone phosphate in the glycolytic pathway. It is an allosteric activator of enzymes such as pyruvate kinase and a substrate used to identify and characterize enzymes such as fructose-1,6-bisphosphate aldolase(s) and fructose-1, 6-bisphosphatase(s). FBP is studied as a neuroprotective agent in brain injury.

Formula: C₆H₁₁Na₃O₁₂P₂
MW: 406.06 g/mol (anhydrous)
Storage Temperature: Freezer
CAS Number: 38099-82-0

Order Now

Specification Test Results

Source/Host Synthetic.
Purity >98% (TLC)
Appearance/Color White to off-white powder.
Solubility Soluble in water.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR