N-Fmoc-N-succinyl-4,7,10-trioxa-1,13-tridecanediamine ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS

CDX-F0204-G001 CDX-F0204-G005
102990-012EA 113.54 USD
102990-012 102990-014
N-Fmoc-N-succinyl-4,7,10-trioxa-1,13-tridecanediamine ≥98% (by HPLC)
N-Fmoc-N-succinyl-4,7,10-trioxa-1,13-tridecanediamine

A bifunctional cross-linker for PEGylation. PEG polymers offer numerous favorable characteristics including high water solubility, high mobility in solution, lack of toxicity and immunogenicity, and ready clearance from the body. Thus, the chemical modification of biologically active compounds, such as peptides, antibody fragments, enzymes, or small molecules with polyethylene glycol chains (referred to as “PEGylation”), is an effective method to tailor molecular properties to particular applications.

Formula: C₂₉H₃₈N₂O₈
Storage Temperature: Freezer
MDL Number: MFCD05663769
CAS Number: 172089-14-4

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (HPLC)
Appearance/Color Colorless crystals.
Solubility Soluble in methanol or acetone.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR