Flumiclorac ≥98% (by HPLC)
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
2-[2-chloro-4-fluoro-5-(1,3,4,5,6,7-hexahydro-1,3-dioxo-2H-isoindol-2-yl)phenoxy]acetic acid
CDX-F0085-M050
CDX-F0085-M250
CDX-F0085-G001
102990-006EA
139.85
USD
102990-006
102990-008
102990-010
Flumiclorac ≥98% (by HPLC)
Flumiclorac
Herbicide. Mode of action is similar to herbicides that inhibit the protoporphyrinogen oxidase (Protox) enzyme in susceptible plant species that leads to an uncontrolled nonenzymatic oxidation of protoporphyrinogen IX (Protogen) to protoporphyrin IX (Proto) and successive necrosis.
Formula:
C₁₆H₁₃ClFNO₅ MW: 353.73 g/mol Storage Temperature: Freezer |
CAS Number:
87547-04-4 |
Specification Test Results
Source/Host | Synthetic |
Purity |
|
Appearance/Color | White powder. |
Solubility | Sligthly soluble in water. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...