Fluorescein octadecyl ester ≥97%

Supplier: ADIPOGEN CORP MS

Synonyms: ETH 7061, Chromoionophore XI

CDX-F0074-M050 CDX-F0074-M500
102989-982EA 171.69 USD
102989-982 102989-984
Fluorescein octadecyl ester ≥97%
Fluorescein octadecyl ester

Chromoionophore XI is used as a lipophilic fluorescent pH indicator.

Formula: C₃₈H₄₈O₅
MW: 584.78 g/mol
Storage Temperature: Refrigerator
CAS Number: 138833-46-2

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 97% (TLC)
Appearance/Color Solid.
Solubility Soluble in chloroform.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR