5(6)-FITC DA ≥97% (by HPLC)
Supplier: ADIPOGEN CORP MS
Synonyms:
Fluorescein diacetate 5(6)-isothiocyanate
CDX-F0029-M010
CDX-F0029-M100
102989-952EA
73.38
USD
102989-952
102989-954
5(6)-FITC DA ≥97% (by HPLC)
5(6)-FITC DA
Fluorescent reagent with high selectivity.
Formula:
C₂₅H₁₅NO₇S MW: 473.45 g/mol Storage Temperature: Freezer |
CAS Number:
871487-69-3 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in chloroform. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...