6-FITC DA 95

Supplier: ADIPOGEN CORP MS

Synonyms: Fluorescein diacetate 6-isothiocyanate

CDX-F0028-M050 CDX-F0028-M250
102989-948EA 213.23 USD
102989-948 102989-950
6-FITC DA 95
6-FITC DA

Fluorescent reagent with high selectivity.

Formula: C₂₅H₁₅NO₇S
MW: 473.45 g/mol
Storage Temperature: Freezer
CAS Number: 890090-49-0

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (HPLC)
Appearance/Color Solid.
Solubility Soluble in chloroform.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR