Fluorescein diacetate-5-maleimide ≥97%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: 5-Maleimido-fluorescein diacetate

CDX-F0018-M025 CDX-F0018-M100
102987-956EA 157.85 USD
102987-956 102989-934
Fluorescein diacetate-5-maleimide ≥97%
Fluorescein diacetate-5-maleimide

Fluorescent marker used extensively in microscopy studies. Can be used in photoluminometric immunoassays for the formation of conjugates with immunoreactants by fluorescent energy transfer.

Formula: C₂₈H₁₇NO₉
MW: 511.4 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00077346
CAS Number: 150322-01-3

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 97% (NMR)
Appearance/Color White powder.
Solubility Soluble in DMSO, dimethylformamide or dioxane.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR