N-Ethanoyl-L-homoserine lactone ≥98%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: Acetyl-L-Homoserine lactone, (S)-N-(2-Oxotetrahydrofuran-3-yl)acetamide, C2-HSL

CDX-E0072-M005 CDX-E0072-M025
102989-924EA 123.23 USD
102989-924 102989-926
N-Ethanoyl-L-homoserine lactone ≥98%
N-Ethanoyl-L-homoserine lactone

N-Ethanoyl-L-homoserine lactone is the shortest alkyl homologue and most polar of a family of mediators of cell to cell interactions in bacterial biofilms. Acylhomoserine lactones have been detected in hundreds of bacterial species. N-acylhomoserine lactones (AHL) are involved in quorum sensing, controlling gene expression, and cellular metabolism. N-Ethanoyl-L-homoserine lactone is not found in nature and can serve as a polar negative control for quorum sensing events.

Formula: C₆H₉NO₃
MW: 143.14 g/mol
Storage Temperature: Freezer
MDL Number: MFCD19160634
CAS Number: 51524-71-1

Order Now

Specification Test Results

Source/Host Synthetic
Purity 98% (NMR)
Appearance/Color White powder.
Solubility Soluble in ethanol, methanol, DMF or DMSO. Poorly soluble in water.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR