N-Ethanoyl-DL-homoserine lactone ≥98% (by NMR)

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: C2-HSL

CDX-E0071-M005 CDX-E0071-M025
102989-920EA 123.23 USD
102989-920 102989-922
N-Ethanoyl-DL-homoserine lactone ≥98% (by NMR)
N-Ethanoyl-DL-homoserine lactone

N-Ethanoyl-DL-homoserine lactone is the shortest alkyl homologue and most polar of a family of mediators of cell to cell interactions in bacterial biofilms. Acylhomoserine lactones have been detected in hundreds of bacterial species. N-acylhomoserine lactones (AHL) are involved in quorum sensing, controlling gene expression, and cellular metabolism. N-Ethanoyl-DL-homoserine lactone is not found in nature and can serve as a polar negative control for quorum sensing events.

Formula: C₆H₉NO₃
MW: 143.14 g/mol
Storage Temperature: Freezer
CAS Number: 114375-78-9

Order Now

Specification Test Results

Source/Host Synthetic
Purity 98% (NMR)
Appearance/Color White to off-white powder.
Solubility Soluble in ethanol, methanol, DMF or DMSO. Poorly soluble in water.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR